Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell InVivoMAb Anti-Mouse CD40L (CD154), MR-1, 25 MG

Catalog No. NC3104675
Encompass_Preferred

The MR-1 monoclonal antibody targets mouse CD154 (CD40 ligand), a 39 kDa accessory molecule crucial for T and B lymphocyte costimulation. It inhibits in vitro B lymphocyte activation by blocking CD154 binding, disrupting germinal center formation, and interfering with T cell responses.

  • Clone: MR-1
  • Reacts with mouse CD154
  • Isotype: Armenian hamster IgG
  • Immunogen: Activated mouse Th1 clone D1.6
  • Reported applications: In vivo/in vitro blocking of CD40/CD40L signaling, Western blot, in vitro Organoids/Organ-on-Chip
  • Formulation: PBS, pH 7.0 (no stabilizers or preservatives)
  • Endotoxin: ≤1 EU/mg
  • Purity: ≥95%
  • Sterility: 0.2 μm filtration
  • Purification: Protein A
  • Molecular Weight: 150 kDa
  • Storage: 4°C, do not freeze

Catalog No. NC3104675 Supplier Bio X Cell Supplier No. BE00171R025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MR-1 monoclonal antibody targets mouse CD154 (CD40 ligand), a 39 kDa accessory molecule crucial for T and B lymphocyte costimulation. It inhibits in vitro B lymphocyte activation by blocking CD154 binding, disrupting germinal center formation, and interfering with T cell responses.

  • Clone: MR-1
  • Reacts with mouse CD154
  • Isotype: Armenian hamster IgG
  • Immunogen: Activated mouse Th1 clone D1.6
  • Reported applications: In vivo/in vitro blocking of CD40/CD40L signaling, Western blot, in vitro Organoids/Organ-on-Chip
  • Formulation: PBS, pH 7.0 (no stabilizers or preservatives)
  • Endotoxin: ≤1 EU/mg
  • Purity: ≥95%
  • Sterility: 0.2 μm filtration
  • Purification: Protein A
  • Molecular Weight: 150 kDa
  • Storage: 4°C, do not freeze

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.