Learn More
Bio X Cell Monoclonal Antibody, anti-mouse CD47, IgG2a, 5 MG
Supplier: Bio X Cell BE0270R005MG
The MIAP301 monoclonal antibody targets mouse CD47 (integrin-associated protein, IAP), a 50 kDa glycosylated transmembrane protein involved in various cellular processes, including immune and angiogenic responses. It neutralizes CD47 both in vivo and in vitro.
- Clone: MIAP301
- Reacts with mouse CD47
- Isotype: Rat IgG2a, κ
- Unconjugated
- Immunogen: Intact CD47 from placenta
- Applications: In vivo/in vitro CD47 blockade, immunofluorescence
- Formulation: PBS, pH 7.0 (no stabilizers/preservatives)
- Endotoxin: ≤1EU/mg
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.