Learn More
Bio X Cell Antibody, Anti-Mouse CD71, Clone 8D3, 100 MG
Supplier: Bio X Cell BE0329R100MG
The 8D3 monoclonal antibody reacts with mouse CD71 (TfR1), a 170-180 kDa type II homodimeric transmembrane glycoprotein. CD71 is found on proliferating cells, reticulocytes, and erythroid precursors, and is essential for iron import. This antibody has been shown to deplete CD71+ cells in vivo.
- Clone: 8D3
- Host species: Rat
- Reactivity: Mouse
- Isotype: Rat IgG2a
- Conjugation: Unconjugated
- Immunogen: Mouse transformed endothelioma cell line t.end1
- Reported applications: In vivo depletion of CD71+ cells, immunofluorescence, IHC (frozen), western blot
- Formulation: PBS, pH 7.0 (no stabilizers or preservatives)
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C. Do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.