Learn More
Bio X Cell Monoclonal Antibody, Ly6C, Rat IgG2a, 25 MG
Supplier: Bio X Cell BE0203R025MG
The Monts 1 monoclonal antibody reacts with mouse Ly6C, a 14-17 kDa member of the Ly-6 superfamily of GPI-anchored cell surface proteins. Ly6C is expressed by monocytes, endothelial cells, granulocytes, and some T cell subsets.
- Clone: Monts 1
- Reactivity: Mouse
- Isotype: Rat IgG2a
- Unconjugated
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/µg)
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 µm filtered
- Production: Purified from cell culture supernatant in an animal-free facility
- Purification: Protein G
- Storage: Store at 4°C, do not freeze
- Molecular weight: 150 kDa
- Reported applications: In vivo macrophage depletion, flow cytometry
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.