Learn More
Bio X Cell anti-mouse MHC Class I Antibody, 5 MG
Supplier: Bio X Cell BE0172R005MG
The Y-3 monoclonal antibody reacts with mouse H-2Kb, H-2Kk, H-2Kq, H-2Kr, H-2Ks, but not H-2Kd MHC class I alloantigens. These heterodimeric molecules present antigenic peptides to CD8 T cells.
- Y-3 clone, mouse reactivity
- Mouse IgG2b, κ isotype, unconjugated
- Immunogen: Con A stimulated spleen cells from BALB.B mice
- Applications: in vivo MHC I blockade, immunopeptidomics, immunoprecipitation, functional assays, flow cytometry
- Formulated in PBS, pH 7.0
- Endotoxin: ≤1EU/mg
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 μm filtration
- Production in animal-free facility; Protein G purification
- Molecular weight: 150 kDa
- Storage: 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.