Learn More
Bio X Cell Anti-NK1.1, Clone PK136, IgG2a, Unconjugated, 50 MG
Supplier: Bio X Cell BE0036R050MG
The PK136 monoclonal antibody reacts with mouse NK1.1 (CD161b/CD161c), a type II integral membrane glycoprotein predominantly expressed on NK cells and NK-T cells. It is involved in NK cell activation, differentiation, IFN-γ production, and cytotoxic granule release, with expression limited to C57BL/6, FVB/N, and NZB mouse strains.
- Clone: PK136
- Reactivities: Mouse
- Isotype: Mouse IgG2a, κ
- Immunogen: Mouse spleen and bone marrow cells (NK1+ enriched)
- Applications: In vivo NK cell depletion, Flow cytometry
- Formulation: PBS, pH 7.0 (no stabilizers/preservatives)
- Endotoxin: ≤1EU/mg
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Production: Purified from animal-free cell culture supernatant
- Purification: Protein A
- Molecular weight: 150 kDa
- Storage: 4°C (do not freeze)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.