Learn More
Bio X Cell Anti-Mouse PD-1, Clone 29F.1A12, IgG2a, 50 MG
Supplier: Bio X Cell BE0273R050MG
The 29F.1A12™ monoclonal antibody targets mouse PD-1 (programmed death-1), also known as CD279, a 50-55 kDa cell surface receptor. It plays key roles in peripheral tolerance and prevention of autoimmune disease, and blocks PD-1 binding to its ligands in vivo.
- Isotype: Rat IgG2a
- Reactivities: Mouse
- Immunogen: Recombinant PD-1-Ig fusion protein
- Purity: ≥95%
- Endotoxin level: ≤1EU/mg (≤0.001EU/μg)
- Sterility: 0.2 μm filtration
- Production: Purified in an animal-free facility
- Storage: Store at stock concentration at 4°C; do not freeze
- Applications: In vivo blocking, Immunohistochemistry, Western blot, Flow cytometry
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.