Learn More
BIOHIPPO InVivoMAb Anti-Mouse VTCN1/B7-H4 Antibody (Iv0257), 100 µg
SDP
Catalog No. 5000860004
Encompass_Preferred
VTCN1/B7-H4 Recombinant Antibody (clone Iv0257) is a recombinant Mouse IgG2a, kappa antibody specific for VTCN1/B7-H4. It reacts with Mouse. Suitable for ELISA, FCM, Neutralization. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.
Catalog No. 50-008-60004 Supplier BIOHIPPO Supplier No. BHA21409566
VTCN1/B7-H4 Recombinant Antibody (clone Iv0257) is a recombinant Mouse IgG2a, kappa antibody specific for VTCN1/B7-H4. It reacts with Mouse. Suitable for ELISA, FCM, Neutralization. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.