Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell InVivoMAb Antibody, Anti-Mouse Galectin-9, RG9-1, 1 MG

Catalog No. NC3239807
Encompass_Preferred

The RG9-1 monoclonal antibody reacts with mouse galectin-9, a 40 kDa S-type lectin involved in innate and adaptive immune responses, including regulation of various T cells and immune cell maturation. It has been reported to block some biological activities of Galectin-9 in vivo.

  • Reacts with mouse galectin-9
  • Host species: Rat
  • Clone: RG9-1
  • Isotype: Rat IgG2b, κ
  • Purification method: Protein G
  • Formulation: PBS, pH 7.0, stabilizer-free
  • Endotoxin: ≤1EU/mg
  • Purity: ≥95%
  • Sterility: 0.2 μm filtered
  • Production: Purified from cell culture supernatant in an animal-free facility
  • Molecular weight: 150 kDa
  • Applications: in vivo Galectin-9 blockade, in vitro Galectin-9 blockade
  • Storage: Store at 4°C, do not freeze

Catalog No. NC3239807 Supplier Bio X Cell Supplier No. BE0218R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The RG9-1 monoclonal antibody reacts with mouse galectin-9, a 40 kDa S-type lectin involved in innate and adaptive immune responses, including regulation of various T cells and immune cell maturation. It has been reported to block some biological activities of Galectin-9 in vivo.

  • Reacts with mouse galectin-9
  • Host species: Rat
  • Clone: RG9-1
  • Isotype: Rat IgG2b, κ
  • Purification method: Protein G
  • Formulation: PBS, pH 7.0, stabilizer-free
  • Endotoxin: ≤1EU/mg
  • Purity: ≥95%
  • Sterility: 0.2 μm filtered
  • Production: Purified from cell culture supernatant in an animal-free facility
  • Molecular weight: 150 kDa
  • Applications: in vivo Galectin-9 blockade, in vitro Galectin-9 blockade
  • Storage: Store at 4°C, do not freeze

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.