Learn More
Bio X Cell InVivoMAb anti-mouse RANKL (CD254), clone IK22/5 - 25 MG (BE0191-R025MG), 25 MG
Supplier: Bio X Cell BE0191R025MG
InVivoMAb anti-mouse RANKL (CD254) is a purified rat monoclonal antibody (clone IK22/5) designed for in vivo blockade of mouse RANKL. Supplied unconjugated in PBS (pH 6.5), the antibody features low endotoxin and high purity to support reliable in vivo experiments.
- Rat monoclonal antibody, clone IK22/5, specific for mouse RANKL (CD254).
- Optimized for in vivo RANKL blockade applications.
- Available in multiple vial sizes, including 25 mg.
- Formulated in PBS, pH 6.5; contains no stabilizers or preservatives.
- Low endotoxin (1 EU/mg) suitable for in vivo use.
- Purity 95% and sterile-filtered (0.2 m).
- Isotype rat IgG2a, ; purified by Protein G.
- Store at 4C; do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.