Learn More
Bio X Cell Bio X Cell, Mouse RANKL IK22/5, Rat IgG2a, 5 MG
Supplier: Bio X Cell BE0191R005MG
The IK22/5 monoclonal antibody reacts with mouse RANKL (CD254), a 35 kDa type II membrane protein involved in regulating T cell and dendritic cell interactions and osteoclast differentiation.
- Isotype: Rat IgG2a, κ
- Immunogen: NSO-derived recombinant mouse RANKL
- Application: in vivo RANKL blockade
- Formulation: PBS, pH 6.5, no stabilizers or preservatives
- Endotoxin: ≤1 EU/mg, determined by LAL assay
- Purity: ≥95%, determined by SDS-PAGE
- Sterility: 0.2 μm filtration
- Production: purified from cell culture supernatant in an animal-free facility
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.