Learn More
Bio X Cell InVivoMAb anti-mouse IFNAR-1, MAR1-5A3, IgG1, 100 MG
Supplier: Bio X Cell BE0241R100MG
The MAR1-5A3 monoclonal antibody reacts with mouse IFNAR-1 (IFN α/β receptor subunit 1), a component of the heterodimeric Type I IFN Receptor complex. It induces cellular responses including anti-viral, anti-microbial, anti-tumor, and autoimmune responses, and inhibits receptor signaling in vitro and in vivo.
- Isotype: Mouse IgG1, κ
- Immunogen: Extracellular domain of mouse IFNAR-1
- Reported applications: IFNAR-1 blockade, Western blot, Flow Cytometry, ELISA
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Production: Purified from animal-free cell culture
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C. Do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.