Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell Anti-Osteopontin, Mouse/Human/Rat SPP1, MPIIIB10, 5 MG

Catalog No. NC3738042
Encompass_Preferred

The MPIIIB10 monoclonal antibody reacts with rat, mouse, and human osteopontin (OPN), also known as SPP1. It is a neutralizing antibody that inhibits tumor growth and synergizes with cell-based immunotherapeutic vaccines in mediating anti-tumor immunity.

  • Clone: MPIIIB10
  • Reactivities: Mouse, human, rat
  • Isotype: Mouse IgG1, κ
  • Conjugation: Unconjugated
  • Immunogen: Rat bone protein fractions
  • Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
  • Endotoxin: ≤1 EU/mg (≤0.001 EU/μg)
  • Purity: ≥95%
  • Sterility: 0.2 μm filtration
  • Purification: Protein G
  • Storage: Store at stock concentration at 4°C. Do not freeze.

Catalog No. NC3738042 Supplier Bio X Cell Supplier No. BE0382R005MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MPIIIB10 monoclonal antibody reacts with rat, mouse, and human osteopontin (OPN), also known as SPP1. It is a neutralizing antibody that inhibits tumor growth and synergizes with cell-based immunotherapeutic vaccines in mediating anti-tumor immunity.

  • Clone: MPIIIB10
  • Reactivities: Mouse, human, rat
  • Isotype: Mouse IgG1, κ
  • Conjugation: Unconjugated
  • Immunogen: Rat bone protein fractions
  • Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
  • Endotoxin: ≤1 EU/mg (≤0.001 EU/μg)
  • Purity: ≥95%
  • Sterility: 0.2 μm filtration
  • Purification: Protein G
  • Storage: Store at stock concentration at 4°C. Do not freeze.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.