Learn More
Bio X Cell InVivoMAb recombinant human IgG1 Fc, 50 MG
Supplier: Bio X Cell BE0096R050MG
This recombinant human IgG1 Fc is the Fc fragment of human IgG1, used as an isotype control for human IgG1 antibodies and fusion proteins containing the human IgG Fc fragment. It has a molecular mass of approximately 34 kDa under reducing conditions.
- Purified from cell culture supernatant in an animal-free facility
- Purification by Protein A
- Formulation in PBS, pH 7.0, without stabilizers or preservatives
- Endotoxin ≤1EU/mg (≤0.001EU/μg)
- Purity ≥95% by SDS-PAGE
- Sterility by 0.2 μm filtration
- Store at stock concentration at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.