Learn More
Bio X Cell Recombinant CTLA-4-Ig, (hum/hum) Clone, 25 MG
Supplier: Bio X Cell BE0099R025MG
This recombinant CTLA-4-Ig fusion protein is purified from cell culture supernatant in an animal-free facility using Protein G. Formulated in PBS, pH 7.0, without stabilizers or preservatives, it boasts an endotoxin level of ≤1EU/mg (≤0.001EU/µg) and ≥95% purity by SDS-PAGE. The solution is sterile, undergoing 0.2 µm filtration.
- Clone: CTLA-4-Ig (hum/hum)
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin level: ≤1EU/mg (≤0.001EU/µg)
- Purity: ≥95% (by SDS-PAGE)
- Sterility: 0.2 µm filtration
- Production: purified from cell culture supernatant in an animal-free facility
- Purification: Protein G
- Storage: store at stock concentration at 4°C. Do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.