Learn More
Bio X Cell Anti-Mouse CD4 Antibody, GK1.5, IgG2b, 50 MG
Supplier: Bio X Cell BP00031R050MG
The GK1.5 monoclonal antibody targets mouse CD4, a 55 kDa cell surface glycoprotein crucial for T cell development and function. This InVivoPlus product is designed for rigorous in vivo research, featuring advanced quality control including ultra-low endotoxin levels and murine pathogen screening.
- Purified via Protein G
- Formulated in PBS, pH 6.5, without stabilizers or preservatives
- Sterile-filtered to 0.2 μm
- Purity ≥95% (SDS-PAGE)
- Endotoxin ≤0.5 EU/mg
- Aggregation <5%
- Applications: in vivo CD4+ T cell depletion, flow cytometry, western blot
- Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.