Learn More
Bio X Cell InVivoPlus Anti-Mouse CD8 Alpha, Clone 53-6.7, Unconjugated, 5 MG
Supplier: Bio X Cell BP00041
The 53-6.7 monoclonal antibody targets mouse CD8α, a transmembrane glycoprotein co-receptor for the T cell receptor (TCR). CD8 is primarily expressed on cytotoxic T cells and is involved in binding to class I MHC molecules. This antibody exhibits depleting activity in vivo.
- Isotype: Rat IgG2a, κ
- Conjugation: Unconjugated
- Immunogen: Mouse spleen cells or thymocyte membranes
- Formulation: PBS, pH 6.5, no stabilizers or preservatives
- Endotoxin: ≤0.5 EU/mg
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Storage: Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.