Learn More
Bio X Cell Monoclonal Antibody, anti-mouse CTLA-4, 9D9, 50 MG
Supplier: Bio X Cell BP0164R050MG
The 9D9 monoclonal antibody reacts with mouse CTLA-4 (CD152), an inhibitory receptor on activated T and B lymphocytes that negatively regulates cell-mediated immune responses. It has been used for blocking/neutralization experiments and intra-tumoral regulatory T cell depletion in tumor models.
- Clone: 9D9
- Reactivities: Mouse
- Isotype: Mouse IgG2b
- Endotoxin: ≤0.5 EU/mg (≤0.0005 EU/μg)
- Aggregation: <5%
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Production: Purified from cell culture supernatant in an animal-free facility
- Purification method: Protein A
- Storage: Store at 4°C; do not freeze
- Applications: In vivo CTLA-4 neutralization, western blot, in vivo intra-tumoral regulatory T cell depletion, in vitro organoids/organ-on-chip
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.