Learn More
Bio X Cell Monoclonal Antibody, 1A8 Clone, Rat IgG2a, Unconjugated, 25 MG
Supplier:  Bio X Cell BP00751R025MG
Encompass_Preferred
This monoclonal antibody targets mouse Ly6G, a 21-25 kDa GPI-anchored cell surface protein involved in cell signaling and adhesion. Ly6G is expressed by myeloid lineage cells, including monocytes, macrophages, granulocytes, and neutrophils. It serves as a reliable cell surface marker for mature granulocytes and peripheral neutrophils, reacting specifically with mouse Ly6G without cross-reactivity to Ly6C.
  • In vivo neutrophil depletion
  • In vivo MDSC depletion
  • Immunofluorescence
  • Immunohistochemistry (paraffin and frozen)
  • Flow cytometry
  • Reacts specifically with mouse Ly6G
  • Unconjugated format
  • High purity (95%)
  • Low endotoxin (0.5 EU/mg)
  • Sterile (0.2 m filtration)
  • Store at 4C; do not freeze
Catalog No. NC1334668