Learn More
Bio X Cell anti-mouse Ly6G, Rat IgG2a, Unconjugated, 5 MG
Supplier: Bio X Cell BP00751R005MG
The 1A8 monoclonal antibody reacts with mouse Ly6G, a 21-25 kDa GPI-anchored cell surface protein expressed by myeloid lineage cells, serving as a marker for mature granulocytes and neutrophils. InVivoPlus™ products feature advanced binding validation, murine pathogen screening, protein aggregation screening, and ultra-low endotoxin levels, ensuring superior quality for in vivo research.
- Clone: 1A8
- Host: Rat; Isotype: Rat IgG2a, κ
- Unconjugated; purified by Protein G
- Formulation: PBS, pH 7.0; no stabilizers or preservatives
- Endotoxin: ≤0.5EU/mg; aggregation: <5%; purity: ≥95%
- Sterility: 0.2 µm filtration
- Applications: In vivo neutrophil depletion, in vivo MDSC depletion, immunofluorescence, immunohistochemistry (paraffin), immunohistochemistry (frozen), flow cytometry
- Storage: Store at stock concentration at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.