Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell Antibody, Ly6G/Ly6C, Clone RB6-8C5, Rat, 50 MG

Catalog No. NC3859706
Encompass_Preferred

The RB6-8C5 monoclonal antibody reacts strongly with mouse Ly6G and weakly with mouse Ly6C (Gr-1), a 21-25 kDa GPI-anchored cell surface protein. Ly6G is expressed by various myeloid lineage cells, including monocytes, macrophages, granulocytes, and neutrophils, serving as a marker for mature granulocytes and peripheral neutrophils. This antibody inhibits the binding of 1A8, which specifically reacts with mouse Ly6G.

  • Endotoxin ≤0.5 EU/mg (≤0.0005 EU/µg)
  • Aggregation <5%
  • Purity ≥95%
  • Sterility by 0.2 µm filtration
  • Host species: Rat
  • Isotype: Rat IgG2b, κ
  • Purification by protein G
  • Formulation in PBS, pH 7.0, without stabilizers or preservatives
  • Store at 4°C, do not freeze

Catalog No. NC3859706 Supplier Bio X Cell Supplier No. BP0075R050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The RB6-8C5 monoclonal antibody reacts strongly with mouse Ly6G and weakly with mouse Ly6C (Gr-1), a 21-25 kDa GPI-anchored cell surface protein. Ly6G is expressed by various myeloid lineage cells, including monocytes, macrophages, granulocytes, and neutrophils, serving as a marker for mature granulocytes and peripheral neutrophils. This antibody inhibits the binding of 1A8, which specifically reacts with mouse Ly6G.

  • Endotoxin ≤0.5 EU/mg (≤0.0005 EU/µg)
  • Aggregation <5%
  • Purity ≥95%
  • Sterility by 0.2 µm filtration
  • Host species: Rat
  • Isotype: Rat IgG2b, κ
  • Purification by protein G
  • Formulation in PBS, pH 7.0, without stabilizers or preservatives
  • Store at 4°C, do not freeze

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.