Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell Anti-Mouse Ly6G/Ly6C Antibody, RB6-8C5, Unconjugated, 100 MG

Catalog No. NC3859707
Encompass_Preferred

The RB6-8C5 monoclonal antibody targets mouse Ly6G and Ly6C (Gr-1), cell surface proteins in the myeloid lineage. Ly6G is a key marker for mature granulocytes and peripheral neutrophils, and the antibody has been shown to inhibit 1A8 antibody binding, which specifically reacts with mouse Ly6G.

  • Reacts with mouse Ly6G and Ly6C
  • Host species: rat
  • Isotype: IgG2b, κ
  • Immunogen: mouse granulocytes
  • Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
  • Endotoxin: ≤0.5 EU/mg; Purity: ≥95%
  • Purification: Protein G
  • Applications: in vivo depletion of Gr-1+ myeloid cells, flow cytometry, immunohistochemistry

Catalog No. NC3859707 Supplier Bio X Cell Supplier No. BP0075R100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The RB6-8C5 monoclonal antibody targets mouse Ly6G and Ly6C (Gr-1), cell surface proteins in the myeloid lineage. Ly6G is a key marker for mature granulocytes and peripheral neutrophils, and the antibody has been shown to inhibit 1A8 antibody binding, which specifically reacts with mouse Ly6G.

  • Reacts with mouse Ly6G and Ly6C
  • Host species: rat
  • Isotype: IgG2b, κ
  • Immunogen: mouse granulocytes
  • Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
  • Endotoxin: ≤0.5 EU/mg; Purity: ≥95%
  • Purification: Protein G
  • Applications: in vivo depletion of Gr-1+ myeloid cells, flow cytometry, immunohistochemistry

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.