Learn More
Bio X Cell Mouse NK1.1, Clone PK136, Unconjugated, 100 MG
Supplier: Bio X Cell BP0036R100MG
The PK136 monoclonal antibody reacts with mouse NK1.1 (CD161b/CD161c, KLRB1, NKR-P1A, Ly-55), a glycoprotein vital for NK cell activation and differentiation. It is expressed on NK cells and NK-T cells, with expression limited to C57BL/6, FVB/N, and NZB mouse strains.
- Isotype: Mouse IgG2a, κ
- Reactivity: Mouse
- Immunogen: Mouse spleen and bone marrow cells
- Formulation: PBS, pH 7.0; no stabilizers
- Endotoxin: ≤0.5 EU/mg (≤0.0005 EU/μg)
- Purity: ≥95% (SDS-PAGE)
- Sterility: 0.2 μm filtration
- Production: Animal-free facility; Protein A purified
- Molecular weight: 150 kDa
- Applications: In vivo NK cell depletion, flow cytometry
- Storage: 4°C stock concentration; do not freeze
- Quality control: Advanced binding validation, murine pathogen screening, aggregation screening, ultra-low endotoxin
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.