Learn More
Bio X Cell anti-mouse PD-1, Clone 29F.1A12, 5 MG
Supplier: Bio X Cell BP0273
The 29F.1A12™ monoclonal antibody targets mouse PD-1 (CD279). It is crucial in research involving peripheral tolerance and autoimmune disease prevention by blocking PD-1 binding to its ligands in vivo.
- Formulation: PBS, pH 7.0, contains no stabilizers or preservatives.
- Endotoxin: ≤0.5EU/mg (≤0.0005EU/μg), determined by LAL assay.
- Aggregation: <5%, determined by SEC.
- Purity: ≥95%, determined by SDS-PAGE.
- Sterility: 0.2 μm filtration.
- Production: purified from cell culture supernatant in an animal-free facility by Protein G purification.
- Molecular weight: 150 kDa.
- Storage: store at stock concentration at 4°C, do not freeze.
- Applications: in vivo blocking of PD-1/PD-L signaling, in vitro PD-1 neutralization, and common immunoassay methods.
- Murine pathogen tests: negative for a comprehensive panel of murine pathogens.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.