Learn More
Bio X Cell Monoclonal Antibody, PD-L1, 10F.9G2, 50 MG
Supplier: Bio X Cell BP0101R050MG
The 10F.9G2™ monoclonal antibody reacts with mouse PD-L1 (programmed death ligand 1), also known as B7-H1 or CD274. This antibody blocks the interaction between PD-L1 and PD-1, and between PD-L1 and B7-1 (CD80), playing a significant role in tumor immune evasion research.
- Murine pathogen screened
- Protein aggregation screened
- Ultra-low endotoxin levels (≤0.5EU/mg)
- Purity ≥95% (SDS-PAGE)
- Purification by protein G
- Formulation: PBS, pH 6.5, no stabilizers or preservatives
- Applications: in vivo PD-L1 blockade, immunofluorescence, flow cytometry, western blot
- Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.