Learn More
Bio X Cell InVivoPlus Control, Mouse IgG1, MOPC-21, 100 MG
Supplier:  Bio X Cell BP0083R100MG
Encompass_Preferred
The MOPC-21 monoclonal antibody is ideal for use as a non-reactive isotype-matched control for mouse IgG1 antibodies in most in vivo and in vitro applications.
  • Contains no stabilizers or preservatives
  • Sterile 0.2 µm filtration
  • Purified from cell culture supernatant in an animal-free facility
  • Advanced binding validation, murine pathogen screening, protein aggregation screening, and ultra-low endotoxin levels ensure superior quality for in vivo research
Catalog No. NC1313978
Add to Cart
Request a Quote