Learn More
Bio X Cell Mouse IgG1 Control, Low Endotoxin, 50 MG
Supplier: Bio X Cell BP0083R050MG
The InVivoPlus mouse IgG1 isotype control is a MOPC-21 monoclonal antibody designed for use as a non-reactive isotype-matched control for mouse IgG1 antibodies in a variety of in vivo and in vitro applications.
- Isotype: mouse IgG1, κ
- Unconjugated format
- Formulation: PBS, pH 6.5, no stabilizers or preservatives
- Endotoxin level ≤0.5 EU/mg
- Aggregation <5%
- Purity ≥95%
- Sterility: 0.2 µm filtration
- Purified from cell culture supernatant in an animal-free facility
- Purification method: protein G
- Molecular weight: 150 kDa
- Recommended storage at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.