Learn More
Bio X Cell Polyclonal Armenian Hamster IgG, Unconjugated, 25 MG
Supplier: Bio X Cell BP0091R025MG
The polyclonal Armenian hamster IgG is purified from Armenian hamster serum. It is ideal for use as a non-reactive control IgG for Armenian hamster antibodies in most in vivo and in vitro applications.
- Isotype: Armenian hamster IgG
- Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
- Endotoxin: ≤0.5EU/mg (≤0.0005EU/μg)
- Aggregation: <5%
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Molecular Weight: 150 kDa
- Storage: Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.