Learn More
Bio X Cell Isotype Control, Rat IgG2a, Anti-Trinitrophenol, 25 MG
Supplier: Bio X Cell BP0089R025MG
This InVivoPlus rat IgG2a isotype control, anti-trinitrophenol, is a monoclonal antibody (clone 2A3) that reacts with trinitrophenol. Since trinitrophenol is not naturally expressed in mammals, this antibody serves as an ideal isotype-matched control for rat IgG2a antibodies in various in vivo and in vitro applications.
- Clone: 2A3
- Isotype: Rat IgG2a, κ
- Conjugation: Unconjugated
- Formulation: PBS, pH 6.5; contains no stabilizers or preservatives
- Endotoxin: ≤0.5 EU/mg (≤0.0005 EU/μg)
- Aggregation: <5%
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C. Do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.