Learn More
Bio X Cell Isotype Control, Rat IgG2a, Anti-trinitrophenol, 5 MG
Supplier: Bio X Cell BP0089R005MG
The InVivoPlus rat IgG2a isotype control, anti-trinitrophenol, is designed for use as an isotype-matched control for rat IgG2a antibodies in both in vivo and in vitro applications.
- Isotype: rat IgG2a, κ
- Clone: 2A3
- Conjugation: unconjugated
- Formulation: PBS, pH 6.5, free of stabilizers or preservatives
- Endotoxin: ≤1 EU/mg, determined by LAL assay
- Purity: ≥95%, determined by SDS-PAGE
- Sterility: 0.2 μm filtration
- Production: purified from cell culture supernatant in an animal-free facility using Protein G
- Storage: store at 4°C; do not freeze
- Molecular weight: 150 kDa
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.