Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell InVivoPure pH 6.0T Dilution Buffer, 50 mL

Catalog No. NC3348409
Encompass_Preferred

InVivoPure™ dilution buffers are specifically formulated and tested to satisfy the stringent requirements for in vivo applications. They are extremely low in endotoxin, have been screened for murine pathogens, tested in animal models for toxicity and are formulated with respect to buffer composition and pH to satisfy the requirements of Bio X Cell's antibodies.

  • Low endotoxin: <0.5 EU/mL (<0.0005 EU/μL)
  • Sterile: 0.2 μM filtered
  • Murine pathogen free: Negative for Norovirus, Parvovirus, Hepatitis Virus, etc.
  • Nontoxic and nonantigenic in animal models
  • Optimized formulation: 5 mM Na2HPO4, 27 mM NaH2PO4, 136 mM NaCl, 0.01% TWEEN® 80
  • Free of undesirable components: No calcium, magnesium, phenol red, or preservatives such as azide
  • For research use only

Catalog No. NC3348409 Supplier Bio X Cell Supplier No. IPT060R050ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

InVivoPure™ dilution buffers are specifically formulated and tested to satisfy the stringent requirements for in vivo applications. They are extremely low in endotoxin, have been screened for murine pathogens, tested in animal models for toxicity and are formulated with respect to buffer composition and pH to satisfy the requirements of Bio X Cell's antibodies.

  • Low endotoxin: <0.5 EU/mL (<0.0005 EU/μL)
  • Sterile: 0.2 μM filtered
  • Murine pathogen free: Negative for Norovirus, Parvovirus, Hepatitis Virus, etc.
  • Nontoxic and nonantigenic in animal models
  • Optimized formulation: 5 mM Na2HPO4, 27 mM NaH2PO4, 136 mM NaCl, 0.01% TWEEN® 80
  • Free of undesirable components: No calcium, magnesium, phenol red, or preservatives such as azide
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.