Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell InVivoPure pH 6.5 Dilution Buffer, 50 mL

Catalog No. NC1415974
Encompass_Preferred

InVivoPure™ dilution buffers are specifically formulated and tested for in vivo applications. They are characterized by extremely low endotoxin levels, screening for murine pathogens, animal model toxicity testing, and buffer composition and pH formulation to meet Bio X Cell's antibody requirements.

  • Endotoxin: ≤ 0.5 EU/mL (≤ 0.0005 EU/μL)
  • Sterility: 0.2 μM filtered
  • Murine pathogen tests: Negative for common murine pathogens
  • Toxicity: Nontoxic and nonantigenic in animal models
  • Concentration: 1X
  • Volume: 50 mL
  • Composition: 16 mM Na2HPO4, 23 mM NaH2PO4, 136 mM NaCl; no calcium, magnesium, phenol red, or preservatives
  • Storage: 4°C
  • Handling: Keep sterile; open only in a biological safety cabinet

Catalog No. NC1415974 Supplier Bio X Cell Supplier No. IP0065R050ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

InVivoPure™ dilution buffers are specifically formulated and tested for in vivo applications. They are characterized by extremely low endotoxin levels, screening for murine pathogens, animal model toxicity testing, and buffer composition and pH formulation to meet Bio X Cell's antibody requirements.

  • Endotoxin: ≤ 0.5 EU/mL (≤ 0.0005 EU/μL)
  • Sterility: 0.2 μM filtered
  • Murine pathogen tests: Negative for common murine pathogens
  • Toxicity: Nontoxic and nonantigenic in animal models
  • Concentration: 1X
  • Volume: 50 mL
  • Composition: 16 mM Na2HPO4, 23 mM NaH2PO4, 136 mM NaCl; no calcium, magnesium, phenol red, or preservatives
  • Storage: 4°C
  • Handling: Keep sterile; open only in a biological safety cabinet

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.