Learn More
Bio X Cell InVivoPure pH 7.0 Dilution Buffer, 50 mL
Catalog No. NC1415979
Encompass_Preferred
InVivoPure™ dilution buffers are specifically formulated and tested to satisfy the stringent requirements for in vivo applications. They are extremely low in endotoxin, have been screened for murine pathogens, tested in animal models for toxicity, and are formulated with respect to buffer composition and pH to satisfy the requirements of Bio X Cell's antibodies.
  • Extremely low in endotoxin (<0.5 EU/mL)
  • Screened for murine pathogens (negative results for all tested)
  • Tested in animal models for toxicity (nontoxic and nonantigenic)
  • Formulated with respect to buffer composition and pH
  • Does not contain calcium, magnesium, phenol red, or preservatives such as azide
  • Sterile (0.2 μM filtered)
Catalog No. NC1415979 Supplier Bio X Cell Supplier No. IP0070R050ML
InVivoPure™ dilution buffers are specifically formulated and tested to satisfy the stringent requirements for in vivo applications. They are extremely low in endotoxin, have been screened for murine pathogens, tested in animal models for toxicity, and are formulated with respect to buffer composition and pH to satisfy the requirements of Bio X Cell's antibodies.
  • Extremely low in endotoxin (<0.5 EU/mL)
  • Screened for murine pathogens (negative results for all tested)
  • Tested in animal models for toxicity (nontoxic and nonantigenic)
  • Formulated with respect to buffer composition and pH
  • Does not contain calcium, magnesium, phenol red, or preservatives such as azide
  • Sterile (0.2 μM filtered)