Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell InVivoSIM Anti-Human PD-L1, IgG1 Lambda, 1 MG

Catalog No. NC3672408
Encompass_Preferred

This InVivoSIM anti-human PD-L1 antibody is an Avelumab biosimilar, provided at 1 mg. It is suitable for in vitro functional assays, PD-L1 blockade, and ELISA applications.

  • Applications include in vitro functional assays, PD-L1 blockade, and ELISA.
  • Antibody host is human.
  • Reacts with human species.
  • Isotype is IgG1, λ.
  • Immunogen is full length human PD-L1.
  • Store solution at 4°C; do not freeze.
  • Ship with adequate blue ice to maintain 4°C.

Catalog No. NC3672408 Supplier Bio X Cell Supplier No. SIM0021R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This InVivoSIM anti-human PD-L1 antibody is an Avelumab biosimilar, provided at 1 mg. It is suitable for in vitro functional assays, PD-L1 blockade, and ELISA applications.

  • Applications include in vitro functional assays, PD-L1 blockade, and ELISA.
  • Antibody host is human.
  • Reacts with human species.
  • Isotype is IgG1, λ.
  • Immunogen is full length human PD-L1.
  • Store solution at 4°C; do not freeze.
  • Ship with adequate blue ice to maintain 4°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.