Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell Anti-human PD-1 Tislelizumab Biosimilar IgG4, 1 MG

Catalog No. NC3865251
Encompass_Preferred

InVivoSIM anti-human PD-1 (Tislelizumab Biosimilar) is a research-grade human IgG4 antibody designed for PD-1 blockade, making it suitable for various research applications.

  • Blocking of PD-1/PD-L signaling
  • Functional assays
  • ELISA
  • Antibody host: human
  • Species reactivity: human
  • Antibody isotype: IgG4, κ
  • Immunogen: human PD-1

Catalog No. NC3865251 Supplier Bio X Cell Supplier No. SIM0038R1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

InVivoSIM anti-human PD-1 (Tislelizumab Biosimilar) is a research-grade human IgG4 antibody designed for PD-1 blockade, making it suitable for various research applications.

  • Blocking of PD-1/PD-L signaling
  • Functional assays
  • ELISA
  • Antibody host: human
  • Species reactivity: human
  • Antibody isotype: IgG4, κ
  • Immunogen: human PD-1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.