Learn More
Electron Microscopy Sciences Invoil Diffusion Pump Oil C 500 ML
SDP
Catalog No. 501898977
Encompass_Preferred
Consists of esters or specially selected hydrocarbon fractions. They were developed for vapor-diffusion pump fluids. They are high purity, very low pressure, and very stable. Pressures lower than 10^-7 torr obtainable. No need for cold trap. Good pumping speed and no corrosive deposits or vapors. Oil C obtains the highest vacuum pressure (10-7 torr) of all of the oils.
Catalog No. 50-189-8977 Supplier Electron Microscopy Sciences Supplier No. 60724
Consists of esters or specially selected hydrocarbon fractions. They were developed for vapor-diffusion pump fluids. They are high purity, very low pressure, and very stable. Pressures lower than 10^-7 torr obtainable. No need for cold trap. Good pumping speed and no corrosive deposits or vapors. Oil C obtains the highest vacuum pressure (10-7 torr) of all of the oils.