Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Inx-sm-56 | 2734878-18-1 | 99.5% | 576.70 | C32H36N2O6S | 1 MG
SDP

Catalog No. 5000203276
Encompass_Preferred

INX-SM-56 is a cytotoxin used as an antibody-drug conjugate (ADC) payload for the synthesis of anti-VISTA antibody conjugates. It is supplied as a white-to-yellow solid with high analytical purity and is intended for research use.

  • Cytotoxin for antibody-drug conjugate synthesis.
  • High purity: 99.47% (analytical).
  • Molecular weight: 576.70; formula: C32H36N2O6S.
  • CAS number: 2734878-18-1.
  • Available in small research sizes such as 1 mg and 5 mg.
  • Storage: 4°C, sealed, away from moisture and light; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-03276 Supplier Medchemexpress LLC Supplier No. HY1471791MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

INX-SM-56 is a cytotoxin used as an antibody-drug conjugate (ADC) payload for the synthesis of anti-VISTA antibody conjugates. It is supplied as a white-to-yellow solid with high analytical purity and is intended for research use.

  • Cytotoxin for antibody-drug conjugate synthesis.
  • High purity: 99.47% (analytical).
  • Molecular weight: 576.70; formula: C32H36N2O6S.
  • CAS number: 2734878-18-1.
  • Available in small research sizes such as 1 mg and 5 mg.
  • Storage: 4°C, sealed, away from moisture and light; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.