Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Inx-sm-56 | 2734878-18-1 | 99.5% | 576.70 g/mol | C32H36N2O6S | 5 MG
SDP

Catalog No. 5000204054
Encompass_Preferred

INX-SM-56 is an ADC cytotoxin intended for use in the synthesis of anti-VISTA antibody-agent conjugates. It is supplied as a solid (white to yellow) with high reported purity, and it has defined solubility and storage profiles suitable for research applications.

  • Cas number 2734878-18-1.
  • Purity 99.5%.
  • Molecular formula C32H36N2O6S.
  • Molecular weight 576.70 g/mol.
  • Form solid, white to yellow.
  • Soluble in DMSO ≥ 50 mg/mL; in vivo formulation ≥ 1.25 mg/mL in 10% DMSO/90% corn oil.
  • Store sealed, away from moisture and light; in solvent: -80°C up to 6 months, -20°C up to 1 month.
  • Available pack sizes include 1 mg and 5 mg.

Catalog No. 50-002-04054 Supplier Medchemexpress LLC Supplier No. HY1471795MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

INX-SM-56 is an ADC cytotoxin intended for use in the synthesis of anti-VISTA antibody-agent conjugates. It is supplied as a solid (white to yellow) with high reported purity, and it has defined solubility and storage profiles suitable for research applications.

  • Cas number 2734878-18-1.
  • Purity 99.5%.
  • Molecular formula C32H36N2O6S.
  • Molecular weight 576.70 g/mol.
  • Form solid, white to yellow.
  • Soluble in DMSO ≥ 50 mg/mL; in vivo formulation ≥ 1.25 mg/mL in 10% DMSO/90% corn oil.
  • Store sealed, away from moisture and light; in solvent: -80°C up to 6 months, -20°C up to 1 month.
  • Available pack sizes include 1 mg and 5 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.