Learn More
Medchemexpress LLC HY-133120 1mg , INY-03-041 CAS:2503017-97-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1331201MG
Encompass_Preferred
HY-133120 1mg INY-03-041 CAS: 2503017-97-6 INY-03-041 is a potent, highly selective and PROTAC-based pan-AKT degrader consisting of the ATP-competitive AKT inhibitor GDC-0068 conjugated to Lenalidomide (Cereblon ligand). INY-03-041 inhibits AKT1, AKT2 and AKT3 with IC50s of 2.0 nM, 6.8 nM and 3.5 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1762