Learn More
Medchemexpress LLC Iny-03-041 | 2503017-97-6 | 99.7% | C44H56ClN7O5 | 10MG
SDP
Catalog No. 5000259611
Encompass_Preferred
INY-03-041 is a PROTAC pan-AKT degrader that links an ATP-competitive AKT inhibitor to a cereblon ligand to induce targeted degradation of AKT isoforms in biochemical and cellular studies. The compound is supplied as a high-purity solid for research use.
  • Potent degradation of AKT1, AKT2, and AKT3 with low-nanomolar activity.
  • Enables investigation of AKT signaling through targeted protein degradation.
  • High purity solid suitable for biochemical and cellular assays.
  • Recommended sealed storage at -20°C; in solvent, store at -80°C for extended stability.
  • Available in small research pack sizes for screening and mechanistic experiments.
Catalog No. 50-002-59611 Supplier Medchemexpress LLC Supplier No. HY13312010MG
INY-03-041 is a PROTAC pan-AKT degrader that links an ATP-competitive AKT inhibitor to a cereblon ligand to induce targeted degradation of AKT isoforms in biochemical and cellular studies. The compound is supplied as a high-purity solid for research use.
  • Potent degradation of AKT1, AKT2, and AKT3 with low-nanomolar activity.
  • Enables investigation of AKT signaling through targeted protein degradation.
  • High purity solid suitable for biochemical and cellular assays.
  • Recommended sealed storage at -20°C; in solvent, store at -80°C for extended stability.
  • Available in small research pack sizes for screening and mechanistic experiments.