Learn More
MSC IO HVAC Controls Thermostat, Tamper-Proof, Heat Only, 65 F
Catalog No. 5000506518
Encompass
The iO HVAC Controls tamper-proof thermostat is designed for heat-only applications, providing precise temperature control at 65°F with 24 Volt operation.
  • Tamper-proof design
  • Heat only operation
  • Fixed temperature control at 65°F
  • Operates on 24 V
  • Amperage of 1 A
  • Outlet mount
  • SPST switch
  • Inside height of 1-1/2 in
  • Inside width of 1/4 in
Catalog No. 50-005-06518 Supplier MSC Supplier No. 45982386
The iO HVAC Controls tamper-proof thermostat is designed for heat-only applications, providing precise temperature control at 65°F with 24 Volt operation.
  • Tamper-proof design
  • Heat only operation
  • Fixed temperature control at 65°F
  • Operates on 24 V
  • Amperage of 1 A
  • Outlet mount
  • SPST switch
  • Inside height of 1-1/2 in
  • Inside width of 1/4 in