Learn More
Hach Company IO9001 Module-includes 1 relay
Supplier:  Hach Company 9494500
Encompass
IO9001 Module (connects through auxiliary port)-includes 1 relay (high voltage)The IO9001 Module adds versatility to the AS950 automatic sampler The IO9001 is available for simple applications that only require 1 high voltage relay For a broader range of inputs and outputs see the IO9004
Catalog No. 50-002-07439