Learn More
Hach Company IO9004 Mod.-incl. multi 4-20mA
Supplier:  Hach Company 9494600
Encompass
IO9004 Module (connects through auxiliary port)-includes multiple 0/4-20 mA outputs and inputsThe IO9004 Module allows the use of anlaog inputs and outputs with the AS950 sampler With the IO9004 module the AS950 provides more input and output options than any other Hach sampler before it
Catalog No. 50-002-07440