Learn More
Bellco Glass, Inc. IOB Impeller Assembly , 500mL-70mm
SDP
Catalog No. 501945050
Encompass_Preferred
Stainless steel shaft design prevents cell damage and minimizes magnetic decoupling. Sealed internal assembly requires less incubator space. For use with bellco 1965-series standard and water jacketed spinner flasks. MADE IN THE USA. Cap size: 70mm Flask size: 500m PADDLE: L X W:2 X 2 IN
Catalog No. 50-194-5050 Supplier Bellco Glass, Inc. Supplier No. 196560015
Stainless steel shaft design prevents cell damage and minimizes magnetic decoupling. Sealed internal assembly requires less incubator space. For use with bellco 1965-series standard and water jacketed spinner flasks. MADE IN THE USA. Cap size: 70mm Flask size: 500m PADDLE: L X W:2 X 2 IN