Learn More
Bellco Glass, Inc. IOB Impeller Assembly , 8L
SDP
Catalog No. 501945054
Encompass_Preferred
Stainless steel shaft design prevents cell damage and minimizes magnetic decoupling. Sealed internal assembly requires less incubator space. For use with bellco 1965-series standard and water jacketed spinner flasks. MADE IN THE USA. Cap size: 100mm Flask size: 8l PADDLE; L X W:6.75 X 5 IN
Catalog No. 50-194-5054 Supplier Bellco Glass, Inc. Supplier No. 196560080
Stainless steel shaft design prevents cell damage and minimizes magnetic decoupling. Sealed internal assembly requires less incubator space. For use with bellco 1965-series standard and water jacketed spinner flasks. MADE IN THE USA. Cap size: 100mm Flask size: 8l PADDLE; L X W:6.75 X 5 IN