Learn More
Bellco Glass, Inc. IOB w/ MicroCarrier Flask, 15L
SDP
Catalog No. 501945065
Encompass_Preferred
Internal impeller decreases risk of contamination. Impeller has an adjustable shaft to align paddle. The large paddle promotes increased oxygenation. MADE IN THE USA. Cap size: 100mm Flask size: 15l Dimensions: 294 x 470mm Sidearm cap: 45mm
Catalog No. 50-194-5065 Supplier Bellco Glass, Inc. Supplier No. 196561150
Internal impeller decreases risk of contamination. Impeller has an adjustable shaft to align paddle. The large paddle promotes increased oxygenation. MADE IN THE USA. Cap size: 100mm Flask size: 15l Dimensions: 294 x 470mm Sidearm cap: 45mm