Learn More
Bellco Glass, Inc. IOB w/ MicroCarrier Flask,36L
SDP
Catalog No. 501945066
Encompass_Preferred
Internal impeller decreases risk of contamination. Impeller has an adjustable shaft to align paddle. The large paddle promotes increased oxygenation. MADE IN THE USA. Cap size: 100mm Flask size: 36l Dimensions: 410 x 545mm Sidearm cap: 45mm
Catalog No. 50-194-5066 Supplier Bellco Glass, Inc. Supplier No. 196561360
Internal impeller decreases risk of contamination. Impeller has an adjustable shaft to align paddle. The large paddle promotes increased oxygenation. MADE IN THE USA. Cap size: 100mm Flask size: 36l Dimensions: 410 x 545mm Sidearm cap: 45mm