Learn More
Bellco Glass, Inc. IOB w/ MicroCarrier Flask, 6L
SDP
Catalog No. 501945063
Encompass_Preferred
Internal impeller decreases risk of contamination. Impeller has an adjustable shaft to align paddle. The large paddle promotes increased oxygenation. MADE IN THE USA. Cap size: 100mm Flask size: 6l Dimensions: 205 x 410mm Sidearm cap: 45mm
Catalog No. 50-194-5063 Supplier Bellco Glass, Inc. Supplier No. 196561060
Internal impeller decreases risk of contamination. Impeller has an adjustable shaft to align paddle. The large paddle promotes increased oxygenation. MADE IN THE USA. Cap size: 100mm Flask size: 6l Dimensions: 205 x 410mm Sidearm cap: 45mm