Learn More
Bellco Glass, Inc. IOB w/ Water Jacketed Flask,250mL
SDP
Catalog No. 501945068
Encompass_Preferred
Internal impeller is useful because it decreases risk of contamination thorugh the cap. Has an adjustable shaft to align the paddle with the bottom of the flask. Large paddle promotes increased oxygenation with low shear. Stainless steel shaft design prevents cell damage and minimizes magnetic decoupling. MADE IN THE USA.
Catalog No. 50-194-5068 Supplier Bellco Glass, Inc. Supplier No. 196562002
Internal impeller is useful because it decreases risk of contamination thorugh the cap. Has an adjustable shaft to align the paddle with the bottom of the flask. Large paddle promotes increased oxygenation with low shear. Stainless steel shaft design prevents cell damage and minimizes magnetic decoupling. MADE IN THE USA.