Learn More
Bellco Glass, Inc. IOB w/ Water Jacketed Flask,3L
SDP
Catalog No. 501945071
Encompass_Preferred
Internal impeller is useful because it decreases risk of contamination thorugh the cap. Has an adjustable shaft to align the paddle with the bottom of the flask. Large paddle promotes increased oxygenation with low shear. Stainless steel shaft design prevents cell damage and minimizes magnetic decoupling. MADE IN THE USA.
Catalog No. 50-194-5071 Supplier Bellco Glass, Inc. Supplier No. 196562030
Internal impeller is useful because it decreases risk of contamination thorugh the cap. Has an adjustable shaft to align the paddle with the bottom of the flask. Large paddle promotes increased oxygenation with low shear. Stainless steel shaft design prevents cell damage and minimizes magnetic decoupling. MADE IN THE USA.